Learn More
Biolegend FITC anti-human CX3CR1
Supplier:  Biolegend 341606
Encompass_Preferred
FITC anti-human CX3CR1 [2A9-1]; Isotype: Rat IgG2b, κ; Reactivity: Human, African Green, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-168-083