Learn More
Biolegend FITC anti-human FPR1
Supplier:  Biolegend 391603
Encompass_Preferred
FITC anti-human FPR1 [W15086B]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-207-8861