Learn More
Biolegend FITC anti-human Ig light chain λ
Supplier:  Biolegend 316606
Encompass_Preferred
FITC anti-human Ig light chain λ [MHL-38]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-166-875