Learn More
Biolegend FITC anti-human IL-1β
Supplier:  Biolegend 508206
Encompass_Preferred
FITC anti-human IL-1β [JK1B-1]; Isotype: Mouse IgG; Reactivity: Human; Apps: ICFC; Size: 100 tests
Catalog No. 50-170-049