Learn More
Biolegend FITC anti-human IL-4
Supplier:  Biolegend 500807
Encompass_Preferred
FITC anti-human IL-4 [MP4-25D2]; Isotype: Rat IgG1, κ; Reactivity: Human, Cross-Reactivity: Swine (Pig, Porcine), Rhesus; Apps: ICFC; Size: 50 ug
Catalog No. 50-169-637