Learn More
Biolegend FITC anti-human IL-8
Supplier:  Biolegend 514604
Encompass_Preferred
FITC anti-human IL-8 [BH0814]; Isotype: Mouse IgG2b, λ; Reactivity: Human; Apps: ICFC; Size: 100 tests
Catalog No. 50-170-183