Learn More
Biolegend FITC anti-human SSEA-4
Supplier:  Biolegend 330410
Encompass_Preferred
FITC anti-human SSEA-4 [MC-813-70]; Isotype: Mouse IgG3, κ; Reactivity: Human, Baboon, Cynomolgus, Mouse, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-167-573