Learn More
Biolegend FITC anti-human TCR Vδ2
Supplier:  Biolegend 331405
Encompass_Preferred
FITC anti-human TCR Vδ2 [B6]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-reactive: Chimpanzee, Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-167-644