Learn More
Biolegend FITC anti-human TCR Vβ5 related subset
Supplier:  Biolegend 316704
Encompass_Preferred
FITC anti-human TCR Vβ5 related subset [MEM-262]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-166-885