Learn More
Biolegend FITC anti-human CD11a/CD18 (LFA-1)
Supplier:  Biolegend 363416
Encompass_Preferred
FITC anti-human CD11a/CD18 (LFA-1) [m24]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-207-8152