Learn More
Biolegend FITC anti-human CD138 (Syndecan-1)
Supplier:  Biolegend 356507
Encompass_Preferred
FITC anti-human CD138 (Syndecan-1) [MI15]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-207-7538