Learn More
Biolegend FITC anti-human CD172a (SIRPα)
Supplier:  Biolegend 372108
Encompass_Preferred
FITC anti-human CD172a (SIRPα) [15-414]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1526516