Learn More
Biolegend FITC anti-human CD19
Supplier:  Biolegend 363007
Encompass_Preferred
FITC anti-human CD19 [SJ25C1]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-207-8111