Learn More
Biolegend FITC anti-human CD236 Glycopho
Catalog No. 502722051
Encompass_Preferred
FITC anti-human CD236 (Glycophorin C) Recombinant [QA20A41]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-272-2051 Supplier Biolegend Supplier No. 378205
FITC anti-human CD236 (Glycophorin C) Recombinant [QA20A41]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests