Learn More
Biolegend FITC anti-human CD27
Catalog No. 502537698
Encompass_Preferred
FITC anti-human CD27 [M-T271]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 500 uL
Catalog No. 50-253-7698 Supplier Biolegend Supplier No. 986910
FITC anti-human CD27 [M-T271]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 500 uL