Learn More
Biolegend FITC anti-human CD304 (Neuropilin-1)
Supplier:  Biolegend 354512
Encompass_Preferred
FITC anti-human CD304 (Neuropilin-1) [12C2]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1449505