Learn More
Biolegend FITC anti-human CD33
Supplier:  Biolegend 366619
Encompass_Preferred
FITC anti-human CD33 [P67.6]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-207-8264