Learn More
Biolegend FITC anti-human CD38
Supplier:  Biolegend 397207
Encompass_Preferred
FITC anti-human CD38 [S17015F]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-207-9297