Learn More
Biolegend FITC anti-human CD38
Supplier:  Biolegend 356609
Encompass_Preferred
FITC anti-human CD38 [HB-7]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-207-7568