Learn More
Cytek Biosciences Inc Antibodies FITC Anti-Human CD45RO (UCHL1) ..... Cat. No. 35-0457-T025 ..... Size 25 tests
Supplier:  Cytek Biosciences Inc 350457T025
Encompass_Preferred
Antibodies FITC Anti-Human CD45RO (UCHL1) ..... Cat. No. 35-0457-T025 ..... Size 25 testsThe UCHL1 antibody reacts with the human CD45 isoform known as CD45RO, a protein tyrosine phosphatase of ‰¥ 220 kDa. CD45 is one of the most abundant hematopoietic markers, and is expressed on all leukocytes (the Leukocyte Common Antigen, LCA). Various isoforms are generated and expressed in cell-specific patterns. With their broad cell distribution, CD45 isoforms are critical for many leukocyte functions, regulating signal transduction and cell activation associated with the T cell receptor, B cell receptor, and IL-2 receptor. Other forms of CD45, with restricted cellular expression, include CD45R (B220), CD45RA and CD45RB.The UCHL1 antibody is widely used as a marker for human CD45RO expression on thymocytes, activated memory T cells, monocytes, macrophages and granulocytes. The antibody is reported to be cross-reactive for Chimpanzee CD45RO.
Catalog No. 50-210-3244