Learn More
Biolegend FITC anti-human CD56 (NCAM) Recombinant
Supplier:  Biolegend 392414
Encompass_Preferred
FITC anti-human CD56 (NCAM) Recombinant [QA17A16]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-207-8904