Learn More
Biolegend FITC anti-human CD8a
Catalog No. 502722511
Encompass_Preferred
FITC anti-human CD8a [RPA-T8]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cynomolgus, Rhesus; Apps: FC; Size: 100 μg
Catalog No. 50-272-2511 Supplier Biolegend Supplier No. 301060
FITC anti-human CD8a [RPA-T8]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cynomolgus, Rhesus; Apps: FC; Size: 100 μg