Learn More
Biolegend FITC anti-human HLA-DQ
Supplier:  Biolegend 361504
Encompass_Preferred
FITC anti-human HLA-DQ [Tü169]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-207-7994