Learn More
Biolegend FITC anti-human IL-22
Supplier:  Biolegend 366712
Encompass_Preferred
FITC anti-human IL-22 [2G12A41]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: ICFC; Size: 100 tests
Catalog No. 50-207-8280