Learn More
Biolegend FITC anti-human β2-microglobulin
Supplier:  Biolegend 395705
Encompass_Preferred
FITC anti-human β2-microglobulin [A17082A]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-207-9191