Learn More
Biolegend FITC anti-mouse CD103 [2E7]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 50 ug
Supplier:  Biolegend 121419
Encompass_Preferred
FITC anti-mouse CD103 [2E7]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-163-938