Learn More
Cytek Biosciences Inc Antibodies FITC Anti-Mouse CD154 (MR1) ..... Cat. No. 35-1541-U025 ..... Size 25 µg
Supplier:  Cytek Biosciences Inc 351541U025
Encompass_Preferred
Antibodies FITC Anti-Mouse CD154 (MR1) ..... Cat. No. 35-1541-U025 ..... Size 25 µgThe MR1 antibody is specific for mouse CD154, a 39 kD glycoprotein that is primarily expressed on activated T cells. CD154 (CD40 ligand, gp39) binds to CD40 expressed on antigen presenting cells and mediates important costimulatory functions between these cells. Signals generated via cognate interactions between CD154-CD40 on activated T and B cells are required for T-dependent B cell responses. The MR1 antibody is reported to block interaction of CD154 with CD40.
Catalog No. 50-210-3262