Learn More
Cytek Biosciences Inc FITC aMu CD16 CD32 Anti
Supplier: Cytek Biosciences Inc 350161U025
Product Name: FITC Anti-Mouse CD16 / CD32 (2.4G2)
Product Class: Antibody
Type: Primary
Product Name: FITC Anti-Mouse CD16 / CD32 (2.4G2)
Cytek Catalog Number: 35-0161-U025
Marker: CD16 / CD32
Target Species: Mouse
Clonality: Monoclonal
Clone: 2.4G2
Host Species: Rat
Isotype: IgG2b
Conjugation: FITC
Size: 25 μg
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.