Learn More
Biolegend FITC anti-mouse CD182 (CXCR2)
Supplier:  Biolegend 149310
Encompass_Preferred
FITC anti-mouse CD182 (CXCR2) [SA044G4]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-207-1851