Learn More
Cytek Biosciences Inc Antibodies FITC Anti-Mouse CD2 (RM2-5) ..... Cat. No. 35-0021-U500 ..... Size 500 µg
Supplier:  Cytek Biosciences Inc 350021U500
Encompass_Preferred
Antibodies FITC Anti-Mouse CD2 (RM2-5) ..... Cat. No. 35-0021-U500 ..... Size 500 µgThe RM2-5 antibody reacts with mouse CD2, an approximately 50 kDa glycoprotein, and a member of the Ig superfamily. CD2, also known as LFA-2, is a receptor for CD48 in the mouse and is expressed on the cell surface of all lymphocytes. CD2 mediates cell to cell adhesion and is involved in T cell activation.RM2-5 is reported to block CD2-mediated cell:cell adhesion. Please choose the appropriate format for each application.
Catalog No. 50-210-3195