Learn More
Cytek Biosciences Inc Antibodies FITC Anti-Mouse CD45.1 (A20) ..... Cat. No. 35-0453-U500 ..... Size 500 µg
Supplier:  Cytek Biosciences Inc 350453U500
Encompass_Preferred
Antibodies FITC Anti-Mouse CD45.1 (A20) ..... Cat. No. 35-0453-U500 ..... Size 500 µgThe A20 antibody reacts with mouse CD45.1, also known as Ly5.1, which is a strain-specific allelic form of the CD45 Leukocyte Common Antigen (LCA). Functionally, CD45 is a protein tyrosine phosphatase whose broad cell distribution supports a critical role in many leukocyte functions, including regulation of signal transduction and cell activation associated with the T cell and B cell receptors.The A20 antibody is typically used as a leukocyte marker in Ly5.1 mouse strains: SJL/J, DA, STS/A and RIII. The antibody has been demonstrated to specific for CD45.1 and is not cross-reactive with CD45.2 bearing cells. 
Catalog No. 50-210-3241