Learn More
Cytek Biosciences Inc Antibodies FITC Anti-Mouse CD5 (53-7.3) ..... Cat. No. 35-0051-U025 ..... Size 25 µg
Supplier:  Cytek Biosciences Inc 350051U025
Encompass_Preferred
Antibodies FITC Anti-Mouse CD5 (53-7.3) ..... Cat. No. 35-0051-U025 ..... Size 25 µgThe 53-7.3 monoclonal antibody is specific for mouse CD5, a 67 kDa protein that is expressed on most thymocytes, mature T cells and the B-1 subset of B cells. CD5 is a member of the scavenger receptor cysteine-rich protein superfamily and is a ligand for the c-type lectin CD72. CD5 is also known as Leu-1 and functions to modulate antigen receptor signaling on T and B cells.
Catalog No. 50-210-3211