Learn More
Biolegend FITC anti-mouse CD64 (FcγRI)
Supplier:  Biolegend 139316
Encompass_Preferred
FITC anti-mouse CD64 (FcγRI) [X54-5/7.1]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. NC1391847