Learn More
Cytek Biosciences Inc Antibodies FITC Anti-Mouse CD80 (B7-1) (16-10A1) ..... Cat. No. 35-0801-U025 ..... Size 25 µg
Supplier:  Cytek Biosciences Inc 350801U025
Encompass_Preferred
Antibodies FITC Anti-Mouse CD80 (B7-1) (16-10A1) ..... Cat. No. 35-0801-U025 ..... Size 25 µgThe 16-10A1 antibody reacts with mouse CD80, also known as B7-1, a 55 kDa type I transmembrane protein ligand for CD152 (CTLA-4) and for CD28, a co-stimulatory receptor for the T cell receptor (TCR). CD28 also binds a second B7 ligand known as CD86 (B7-2). Both CD80 and CD86 are expressed on activated B cells and antigen-presenting cells. These ligands trigger CD28 signaling in concert with TCR activation to drive T cell proliferation, induce high-level expression of IL-2, impart resistance to apoptosis, and enhance T cell cytotoxicity. The interaction / co-stimulatory signaling between the B7 ligands and CD28 or CTLA-4 provides crucial communication between T cells and B cells or APCs to coordinate the adaptive immune response.
Catalog No. 50-210-3253