Learn More
Cytek Biosciences Inc FITC aMu MHCII Anti Primary
Supplier: Cytek Biosciences Inc 355321U025
Product Name: FITC Anti-Mouse MHC Class II (I-A/I-E) (M5/114.15.2)
Product Class: Antibody
Type: Primary
Product Name: FITC Anti-Mouse MHC Class II (I-A/I-E) (M5/114.15.2)
Cytek Catalog Number: 35-5321-U025
Marker: MHC Class II (I-A/I-E)
Target Species: Mouse
Clonality: Monoclonal
Clone: M5/114.15.2
Host Species: Rat
Isotype: IgG2b, k
Conjugation: FITC
Size: 25 μg
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.