Learn More
Biolegend FITC anti-mouse NK-1.1
Supplier:  Biolegend 156507
Encompass_Preferred
FITC anti-mouse NK-1.1 [S17016D]; Isotype: Mouse IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-207-2293