Learn More
Biolegend FITC anti-mouse/rat XCR1
Supplier:  Biolegend 148209
Encompass_Preferred
FITC anti-mouse/rat XCR1 [ZET]; Isotype: Mouse IgG2b, κ; Reactivity: Mouse, Rat; Apps: FC; Size: 25 ug
Catalog No. 50-207-1783