Learn More
Biolegend FITC anti-mouse CD102
Supplier:  Biolegend 105606
Encompass_Preferred
FITC anti-mouse CD102 [3C4 (MIC2/4)]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 500 ug
Catalog No. 50-163-063