Learn More
Biolegend FITC anti-mouse CD104
Supplier:  Biolegend 123605
Encompass_Preferred
FITC anti-mouse CD104 [346-11A]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-164-131