Learn More
Biolegend FITC anti-mouse CD107b (Mac-3)
Supplier:  Biolegend 108504
Encompass_Preferred
FITC anti-mouse CD107b (Mac-3) [M3/84]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 500 ug
Catalog No. 50-163-270