Learn More
Biolegend FITC anti-mouse CD170 (Siglec-F)
Supplier:  Biolegend 155504
Encompass_Preferred
FITC anti-mouse CD170 (Siglec-F) [S17007L]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-207-2205