Learn More
Biolegend FITC anti-mouse CD18
Supplier:  Biolegend 101405
Encompass_Preferred
FITC anti-mouse CD18 [M18/2]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-162-559