Learn More
Biolegend FITC anti-mouse CD183 (CXCR3)
Supplier:  Biolegend 126536
Encompass_Preferred
FITC anti-mouse CD183 (CXCR3) [CXCR3-173]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-207-0721