Learn More
Biolegend FITC anti-mouse CD1d (CD1.1, Ly-38)
Supplier:  Biolegend 123507
Encompass_Preferred
FITC anti-mouse CD1d (CD1.1, Ly-38) [1B1]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-164-119