Learn More
Biolegend FITC anti-mouse CD200R (OX2R)
Supplier:  Biolegend 123909
Encompass_Preferred
FITC anti-mouse CD200R (OX2R) [OX-110]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-164-148