Learn More
Biolegend FITC anti-mouse CD28
Supplier:  Biolegend 122008
Encompass_Preferred
FITC anti-mouse CD28 [E18]; Isotype: Mouse IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 500 ug
Catalog No. 50-163-999