Learn More
Biolegend FITC anti-mouse CD31
Supplier:  Biolegend 102406
Encompass_Preferred
FITC anti-mouse CD31 [390]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 500 ug
Catalog No. 50-162-679