Learn More
Biolegend FITC anti-mouse CD326 (Ep-CAM)
Supplier:  Biolegend 118208
Encompass_Preferred
FITC anti-mouse CD326 (Ep-CAM) [G8.8]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 500 ug
Catalog No. 50-163-757