Learn More
Biolegend FITC anti-mouse CD38
Supplier:  Biolegend 102705
Encompass_Preferred
FITC anti-mouse CD38 [90]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-162-721