Learn More
Biolegend FITC anti-mouse CD4
Supplier:  Biolegend 116004
Encompass_Preferred
FITC anti-mouse CD4 [RM4-4]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 500 ug
Catalog No. 50-163-598